missing translation for 'onlineSavingsMsg'
Learn More

S-Ethyl Thioacetate 98.0+%, TCI America™

Catalog No. E070825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E070825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E070825G Supplier TCI America Supplier No. E070825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-60-5
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00015178
InChI Key APTGPWJUOYMUCE-UHFFFAOYSA-N
Synonym s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester
PubChem CID 61171
IUPAC Name S-ethyl ethanethioate
SMILES CCSC(=O)C

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 g
UN Number 3272
Formula Weight 104.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Ethyl Thioacetate