missing translation for 'onlineSavingsMsg'
Learn More

(S)-Epichlorohydrin 98.0+%, TCI America™

Supplier:  TCI America E053325G

 View more versions of this product

Catalog No. E053325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-Epichlorohydrin
Yellow
C3H5ClO
25 g
s-+-epichlorohydrin, s-epichlorohydrin, 2s-2-chloromethyl oxirane, s-chloromethyl oxirane, s-3-chloropropylene oxide, s-2-chloromethyl oxirane, epichlorohydrin, +, unii-scr89b4r6o, ccris 6388
ClC[C@@H]1CO1
92.52
CHEBI:37145
≥98.0% (GC)
67843-74-7
112°C
MFCD00077760
2023
BRLQWZUYTZBJKN-GSVOUGTGSA-N
(2S)-2-(chloromethyl)oxirane
149428
92.52
Liquid

Chemical Identifiers

67843-74-7
92.52
BRLQWZUYTZBJKN-GSVOUGTGSA-N
149428
(2S)-2-(chloromethyl)oxirane
C3H5ClO
MFCD00077760
s-+-epichlorohydrin, s-epichlorohydrin, 2s-2-chloromethyl oxirane, s-chloromethyl oxirane, s-3-chloropropylene oxide, s-2-chloromethyl oxirane, epichlorohydrin, +, unii-scr89b4r6o, ccris 6388
CHEBI:37145
ClC[C@@H]1CO1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0275

RTECSNumber : RR0427100

TSCA : No

SDS
Documents

Documents

Promotions

Promotions