missing translation for 'onlineSavingsMsg'
Learn More

(S)-Epichlorohydrin 98.0+%, TCI America™

Catalog No. E05335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05335G 5 g
E053325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05335G Supplier TCI America Supplier No. E05335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67843-74-7
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077760
InChI Key BRLQWZUYTZBJKN-GSVOUGTGSA-N
Synonym s-+-epichlorohydrin, s-epichlorohydrin, 2s-2-chloromethyl oxirane, s-chloromethyl oxirane, s-3-chloropropylene oxide, s-2-chloromethyl oxirane, epichlorohydrin, +, unii-scr89b4r6o, ccris 6388
PubChem CID 149428
ChEBI CHEBI:37145
IUPAC Name (2S)-2-(chloromethyl)oxirane
SMILES ClC[C@@H]1CO1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
UN Number 2023
Formula Weight 92.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Epichlorohydrin
EINECSNumber (2)-0275
RTECSNumber RR0427100
TSCA No