missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-DBD-APy 98.0+%, TCI America™
[HPLC Labeling Reagent for e.e.Determination]
Supplier: TCI America A5560100MG
Specifications
| (S)-(+)-DBD-APy [=(S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)- | |
| 130°C | |
| MFCD00191378 | |
| (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole | |
| CN(C)S(=O)(=O)C1=CC=C(N2CCC(N)C2)C2=NON=C12 | |
| 311.36 | |
| 311.36 | |
| Crystalline Powder |
| 143112-50-9 | |
| C12H17N5O3S | |
| 100 mg | |
| SZJGHUNJZRXXCA-UHFFFAOYNA-N | |
| 7-(3-aminopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide | |
| 15682839 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 143112-50-9 | |
| 311.36 | |
| SZJGHUNJZRXXCA-UHFFFAOYNA-N | |
| 15682839 | |
| CN(C)S(=O)(=O)C1=CC=C(N2CCC(N)C2)C2=NON=C12 |
| C12H17N5O3S | |
| MFCD00191378 | |
| (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-(N,N-Dimethylaminosulfonyl)-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole | |
| 7-(3-aminopyrrolidin-1-yl)-N,N-dimethyl-2,1,3-benzoxadiazole-4-sulfonamide |
Safety and Handling
TSCA : No