missing translation for 'onlineSavingsMsg'
Learn More

S-Curve™ X-Large Storage Bin Dispenser

Catalog No. 19162438
Change view
Click to view available options
Dimensions (L x W x H):
18 x 16 x 10 in. (WxHxD)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
19162438 18 x 16 x 10 in. (WxHxD)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19162438 Supplier S-Curve™ Supplier No. SBD18PETG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Heavy duty wall mount bracket

Hold and dispenses a large volume of items without spilling

Designed for bulk items such as gloves, bouffants, shoe covers, and beard covers.
  • 1/4 in. clear PETG construction
  • Easy-access front opening
  • Wall mounts or free standing
  • Flame treated edges for smoothness
  • Full length hinged top for greater durability
  • Special design prevents items from falling out
Comes packaged in a poly bag.

Specifications

Color Clear
Material PETG
Mounting Type Wall or Free Standing
Includes Full-length hinged lid
Dimensions (L x W x H) 18 x 16 x 10 in. (WxHxD)
Thickness 0.25 in.