missing translation for 'onlineSavingsMsg'
Learn More

S-Curve™ Safety Glass Dispenser

Catalog No. 19170657
Change view
Click to view available options
Material:
PETG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19170657 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19170657 Supplier S-Curve™ Supplier No. SGD07PETG

missing translation for 'validMainframeMsg'

Specifications

Color Clear
Product Type Safety Glasses Dispenser
Dimensions (L x W x H) 7 x 24.5 x 5 in.
For Use With (Application) Holds up to 7 pairs of Glasses
Material PETG
Mounting Wall Mount