missing translation for 'onlineSavingsMsg'
Learn More

S-Curve™ Safety Glass Holder with Foam Pad

Catalog No. 19999940
Change view
Click to view available options
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19999940 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19999940 Supplier S-Curve™ Supplier No. SGH001

Includes

Wall anchors and screws for wall mounting.

Wall-mount or freestanding

  • Keep safety glasses accessible and scratch-free
  • Lightweight, clear acrylic case with a foam pad on the bottom to protect glasses from scratching

Specifications

Description Safety Glass Holder
Color Clear
Product Type Eyewear Storage Box
For Use With (Application) Keep safety glasses accessible and scratch-free
Includes Wall anchors and screws for wall mounting
Material Acrylic