missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-α-Amino-γ-butyrolactone hydrobromide, 99%, Thermo Scientific Chemicals

Catalog No. AC259940050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259940050 5 g Glass bottle
AC259940010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC259940050 Supplier Thermo Scientific Chemicals Supplier No. 259940050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15295-77-9
Molecular Formula C4H8BrNO2
Molecular Weight (g/mol) 182.02
MDL Number MFCD00012723,MFCD00674493
InChI Key MKLNTBLOABOJFZ-UHFFFAOYNA-N
Synonym s-3-aminodihydrofuran-2 3h-one hydrobromide, s---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s-2-amino-4-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, s---alpha-amino-gamma-butyrolactonehydrobromide, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr
PubChem CID 16213138
SMILES Br.NC1CCOC1=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 236.0°C to 238.9°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 21.50 (20.00°C c=4,water)
Specific Rotation − 21.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone
IUPAC Name 3-aminooxolan-2-one hydrobromide
Formula Weight 182.02
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-Amino-γ-butyrolactone hydrobromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only