missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-alpha-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™

Catalog No. P13681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13681G Supplier TCI America Supplier No. P13681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 340-06-7
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.138
MDL Number MFCD00077845
InChI Key VNOMEAQPOMDWSR-ZETCQYMHSA-N
Synonym s-+-alpha-trifluoromethyl benzyl alcohol, s-2,2,2-trifluoro-1-phenylethanol, 1s-2,2,2-trifluoro-1-phenylethan-1-ol, 1s-2,2,2-trifluoro-1-phenylethanol, s-1-phenyl-2,2,2-trifluoroethanol, s-+-1-phenyl-2,2,2-trifluoroethanol, pubchem15591, s-1-phenyltrifluoroethanol, s-a-trifluoromethyl benzyl alcohol, 1s-1-phenyl-2,2,2-trifluoroethanol
PubChem CID 1472401
IUPAC Name (1S)-2,2,2-trifluoro-1-phenylethanol
SMILES C1=CC=C(C=C1)C(C(F)(F)F)O

Specifications

Color Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 176.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-alpha-(Trifluoromethyl)benzyl Alcohol