missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-alpha-Methylbenzyl Isocyanate 98.0+%, TCI America™

Catalog No. I03351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03351G 1 g
I03355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03351G Supplier TCI America Supplier No. I03351G

Chemical Identifiers

CAS 14649-03-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00002036
InChI Key JJSCUXAFAJEQGB-QMMMGPOBSA-N
Synonym s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
PubChem CID 2724173
IUPAC Name [(1S)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 1 g
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-alpha-Methylbenzyl Isocyanate
TSCA No
Recommended Storage Refrigerator