missing translation for 'onlineSavingsMsg'
Learn More

S-Acetylthiocholine iodide, 98%

CAS: 1866-15-5 | C7H16INOS | 289.175 g/mol

$54.73 - $598.44

Chemical Identifiers

CAS 1866-15-5
Molecular Formula C7H16INOS
Molecular Weight (g/mol) 289.175
MDL Number MFCD00011819
InChI Key NTBLZMAMTZXLBP-UHFFFAOYSA-M
Synonym acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
PubChem CID 74629
IUPAC Name 2-acetylsulfanylethyl(trimethyl)azanium;iodide
SMILES CC(=O)SCC[N+](C)(C)C.[I-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1680203
View Documents
Thermo Scientific Chemicals
A1680203
1 g
Each for $54.73
Only null left
 
AAA1680206
View Documents
Thermo Scientific Chemicals
A1680206
5 g
Each for $161.70
Only null left
 
AAA1680214
View Documents
Thermo Scientific Chemicals
A1680214
25 g
Each for $598.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1866-15-5
289.175
NTBLZMAMTZXLBP-UHFFFAOYSA-M
74629
CC(=O)SCC[N+](C)(C)C.[I-]
C7H16INOS
MFCD00011819
acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
2-acetylsulfanylethyl(trimethyl)azanium;iodide

Specifications

1866-15-5
C7H16INOS
1 g
3916578
acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
CC(=O)SCC[N+](C)(C)C.[I-]
289.175
289.18
S-Acetylthiocholine iodide
205°C to 209°C
MFCD00011819
UN2811
Hygroscopic; Light sensitive
NTBLZMAMTZXLBP-UHFFFAOYSA-M
2-acetylsulfanylethyl(trimethyl)azanium;iodide
74629
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 217-474-1

RTECSNumber : FZ9865000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only