missing translation for 'onlineSavingsMsg'
Learn More

S-Acetylthiocholine iodide, 98%

Catalog No. AC102710250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102710250 25 g Glass bottle
AC102710050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102710250 Supplier Thermo Scientific Chemicals Supplier No. 102710250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1866-15-5 | C7H16INOS | 289.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1866-15-5
MDL Number MFCD00011819
InChI Key NTBLZMAMTZXLBP-UHFFFAOYSA-M
Synonym acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
PubChem CID 74629
IUPAC Name 2-acetylsulfanylethyl(trimethyl)azanium;iodide
SMILES CC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 205°C to 210°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C7H16INOS
Linear Formula CH3COSCH2CH2N(CH3)3I
Quantity 25 g
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol
Molecular Weight (g/mol) 289.17
Formula Weight 289.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material S-Acetylthiocholine iodide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
EINECSNumber 217-474-1
RTECSNumber FZ9865000
TSCA TSCA

RUO – Research Use Only