missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-6-Methyl-5-hepten-2-ol, 99%

Catalog No. p-7047350
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL18884MD 250 mg
AAL1888403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL18884MD Supplier Thermo Scientific Chemicals Supplier No. L18884MD
Only null left

CAS: 58917-26-3 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58917-26-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD03093080
InChI Key OHEFFKYYKJVVOX-QMMMGPOBSA-N
Synonym s-sulcatol, s-6-methylhept-5-en-2-ol, 2s-6-methylhept-5-en-2-ol, 2s-6-methyl-5-hepten-2-ol, s-+-6-methyl-5-hepten-2-ol, 6-methyl-5-hepten-2s-ol, 5-hepten-2-ol, 6-methyl-, 2s, +-sulcatol, s-6-methyl-hept-5-en-2-ol
PubChem CID 6971127
IUPAC Name (2S)-6-methylhept-5-en-2-ol
SMILES CC(CCC=C(C)C)O

Specifications

Density 0.844
Boiling Point 78°C to 80°C (15 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.448
Quantity 250 mg
Beilstein 1720071
Optical Rotation +18° (Neat)
Formula Weight 128.22
Percent Purity 99%
Chemical Name or Material (S)-(+)-6-Methyl-5-hepten-2-ol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only