missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-6-Methyl-1-octanol 98.0+%, TCI America™

Catalog No. p-7132466
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09661ML 1 mL
M09665ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09661ML Supplier TCI America Supplier No. M09661ML

Chemical Identifiers

CAS 110453-78-6
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00221493
InChI Key WWRGKAMABZHMCN-UHFFFAOYNA-N
PubChem CID 13548104
ChEBI CHEBI:83010
IUPAC Name 6-methyloctan-1-ol
SMILES CCC(C)CCCCCO

Specifications

Color Colorless
Boiling Point 208°C
Quantity 1 mL
Formula Weight 144.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-6-Methyl-1-octanol