missing translation for 'onlineSavingsMsg'
Learn More

(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. p-7125671
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28321G 1 g
C28325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28321G Supplier TCI America Supplier No. C28321G
Only null left

Chemical Identifiers

CAS 160982-16-1
Molecular Formula C6H6ClNO3S2
Molecular Weight (g/mol) 239.688
MDL Number MFCD09033311
InChI Key OFJGKGNJDCLNPM-SCSAIBSYSA-N
PubChem CID 11746721
IUPAC Name (4S)-6-chloro-1,1-dioxo-3,4-dihydro-2H-thieno[3,2-e]thiazin-4-ol
SMILES C1C(C2=C(SC(=C2)Cl)S(=O)(=O)N1)O

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Formula Weight 239.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine 1,1-Dioxide