missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-6,7-Diméthoxy-1,2,3,4-tétrahydroisoquinoline-3-carbooxylique Chlorhydrate d’acide 98,0+%, TCI America™

Numéro de catalogue. d43755g
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D43755G 5 g
D43751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D43755G Fournisseur TCI America Code fournisseur D43755G

Identifiants chimiques

CAS 82586-62-7
Molecular Formula C12H16ClNO4
Molecular Weight (g/mol) 273.713
MDL Number MFCD01861226
InChI Key ROWPWZMWICGKBY-FVGYRXGTSA-N
PubChem CID 2734261
IUPAC Name (3S)-6,7-diméthoxy-1,2,3,4-tétrahydroisoquinoline-3-acide carboxylique; Chlorhydrate
SMILES COC1=C(C=C2CNC(CC2=C1)C(=O)O)OC.Cl

Spécifications

Melting Point 272°C
Color Blanc
Quantity 5 g
Formula Weight 273.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material (S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid Hydrochloride