missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. D43751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43751G 1 g
D43755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43751G Supplier TCI America Supplier No. D43751G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82586-62-7
Molecular Formula C12H16ClNO4
Molecular Weight (g/mol) 273.713
MDL Number MFCD01861226
InChI Key ROWPWZMWICGKBY-FVGYRXGTSA-N
PubChem CID 2734261
IUPAC Name (3S)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid;hydrochloride
SMILES COC1=C(C=C2CNC(CC2=C1)C(=O)O)OC.Cl

Specifications

Melting Point 272°C
Color White
Quantity 1 g
Formula Weight 273.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid Hydrochloride