missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. O02761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02761G 1 g
O02765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02761G Supplier TCI America Supplier No. O02761G

Chemical Identifiers

CAS 21461-84-7
Molecular Formula C5H5O4
Molecular Weight (g/mol) 129.09
MDL Number MFCD00064516
InChI Key QVADRSWDTZDDGR-VKHMYHEASA-M
Synonym s-5-oxotetrahydrofuran-2-carboxylic acid, 2s-5-oxooxolane-2-carboxylic acid, s-+-5-oxo-2-tetrahydrofurancarboxylic acid, s-+-5-oxotetrahydrofuran-2-carboxylic acid, s-5-oxo-tetrahydrofuran-2-carboxylic acid, s-+-5-oxo-2-tetrahydrofuroic acid, s-5-oxotetrahydrofuran-2-carboxylicacid, 2s-5-oxotetrahydrofuran-2-carboxylic acid, s-alpha-hydroxyglutaric acid-gamma-lactone, s-tetrahydro-5-oxo-2-furancarboxylic acid
PubChem CID 636468
ChEBI CHEBI:73677
IUPAC Name (2S)-5-oxooxolane-2-carboxylate
SMILES [O-]C(=O)[C@@H]1CCC(=O)O1

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 130.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-5-Oxotetrahydrofuran-2-carboxylic Acid