missing translation for 'onlineSavingsMsg'
Learn More

(S)-5-(Hydroxymethyl)-2-pyrrolidinone 98.0+%, TCI America™

Catalog No. p-7130772
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08671G 1 g
H08675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08671G Supplier TCI America Supplier No. H08671G

Chemical Identifiers

CAS 17342-08-4
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077792
InChI Key HOBJEFOCIRXQKH-BYPYZUCNSA-N
Synonym l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847
PubChem CID 643511
IUPAC Name (5S)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 86°C
Quantity 1 g
Formula Weight 115.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-5-(Hydroxymethyl)-2-pyrrolidinone