missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone, 98%

Catalog No. AAL1835906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1835906 5 g
AAL1835903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1835906 Supplier Thermo Scientific Chemicals Supplier No. L1835906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17342-08-4 | C5H9NO2 | 115.132 g/mol

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone serves as a building block in the synthesis of (R)- and (S)-diaminovaleric acids1 and of 5-azasemicorrin bidentate nitrogen ligands for enantioselective catalysis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 17342-08-4
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077792
InChI Key HOBJEFOCIRXQKH-BYPYZUCNSA-N
Synonym l-pyroglutaminol, s-+-5-hydroxymethyl-2-pyrrolidinone, 5s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl pyrrolidin-2-one, s-5-hydroxymethyl-2-pyrrolidinone, 2-pyrrolidinone, 5-hydroxymethyl-, 5s, s-pyroglutaminol, l-pyroglutamol, pubchem13847
PubChem CID 643511
IUPAC Name (5S)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 78°C to 82°C
Boiling Point 147°C to 149°C (0.06 mmHg)
Quantity 5 g
Beilstein 4657914
Solubility Information Soluble in water.
Optical Rotation +30° (c=2 in Ethanol)
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only