missing translation for 'onlineSavingsMsg'
Learn More

(S)-[4-(Pyridin-4-yl)-4,5-dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepine-2,6-diyl]bis[bis[4-(tert-butyl)phenyl- 85+%, TCI America™

Catalog No. P238050MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P238050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P238050MG Supplier TCI America Supplier No. P238050MG
Only null left

Chemical Identifiers

CAS 1883396-49-3
Molecular Formula C69H72N2O2

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 961.35
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-[4-(Pyridin-4-yl)-4,5-dihydro-3H-dinaphtho[2,1-c:1′,2′-e]azepine-2,6-diyl]bis[bis[4-