missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Phenylthiazolidine-2-thione 98.0+%, TCI America™

Catalog No. P19601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19601G Supplier TCI America Supplier No. P19601G
Only null left

Chemical Identifiers

CAS 185137-29-5
Molecular Formula C9H9NS2
Molecular Weight (g/mol) 195.298
MDL Number MFCD06658214
InChI Key IEXSISKCCADMLK-MRVPVSSYSA-N
PubChem CID 11333042
IUPAC Name (4S)-4-phenyl-1,3-thiazolidine-2-thione
SMILES C1C(NC(=S)S1)C2=CC=CC=C2

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 1 g
Formula Weight 195.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-Phenylthiazolidine-2-thione