missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Penten-2-ol, 97%

Catalog No. AAB2572806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2572806 5 g
AAB2572803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2572806 Supplier Thermo Scientific Chemicals Supplier No. B2572806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55563-79-6 | C5H10O | 86.134 g/mol

(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-4-Penten-2-ol is involved in the synthesis of goniothalamin, hexadecanolide, massoia lactone, and parasorbic acid through sequential allylboration-esterification ring-closing metathesis reactions. It is also used to prepare 2-pentanone.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 55563-79-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD03427200
InChI Key ZHZCYWWNFQUZOR-YFKPBYRVSA-N
Synonym s-+-4-penten-2-ol, 4-penten-2-ol, 2s, 2s-pent-4-en-2-ol, 2s-4penten-2-ol, s-4-pentene-2-ol, 2s-4-penten-2-ol, s-pent-4-en-2-ol, s-2-hydroxypent-4-ene
PubChem CID 6994332
IUPAC Name (2S)-pent-4-en-2-ol
SMILES CC(CC=C)O

Specifications

Density 0.837
Boiling Point 115°C to 116°C
Flash Point 30°C (86°F)
Refractive Index 1.425
Quantity 5 g
UN Number UN1987
Optical Rotation +5.2° (c=1 in Chloroform)
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material (S)-(+)-4-Penten-2-ol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only