missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Methyl-1-hexanol 98.0+%, TCI America™

Catalog No. p-7132465
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09641ML 1 mL
M09645ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09641ML Supplier TCI America Supplier No. M09641ML

Chemical Identifiers

CAS 1767-46-0
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00221492
InChI Key YNPVNLWKVZZBTM-ZETCQYMHSA-N
PubChem CID 13463446
IUPAC Name (4S)-4-methylhexan-1-ol
SMILES CCC(C)CCCO

Specifications

Color Yellow
Boiling Point 126°C
Quantity 1 mL
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-4-Methyl-1-hexanol