missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone 97.0+%, TCI America™

$80.16 - $234.87

Chemical Identifiers

CAS 77877-19-1
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD00043401
InChI Key HOWPHXVPNNPSAZ-SSDOTTSWSA-N
PubChem CID 853160
IUPAC Name (4S)-3-propanoyl-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1C(COC1=O)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05731G
View Documents
TCI America
I05731G
1 g
Each for $80.16
Only null left
 
I05735G
View Documents
TCI America
I05735G
5 g
Each for $234.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

77877-19-1
185.223
HOWPHXVPNNPSAZ-SSDOTTSWSA-N
(4S)-3-propanoyl-4-propan-2-yl-1,3-oxazolidin-2-one
C9H15NO3
MFCD00043401
853160
CCC(=O)N1C(COC1=O)C(C)C

Specifications

77877-19-1
C9H15NO3
1 g
CCC(=O)N1C(COC1=O)C(C)C
185.223
185.22
Liquid
Yellow
MFCD00043401
HOWPHXVPNNPSAZ-SSDOTTSWSA-N
(4S)-3-propanoyl-4-propan-2-yl-1,3-oxazolidin-2-one
853160
≥97.0% (GC)
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'