missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 185.22 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone |
Safety and Handling
| TSCA | No |