missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Hydroxy-2-pyrrolidone 98.0+%, TCI America™

Catalog No. H13681G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13681G Supplier TCI America Supplier No. H13681G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68108-18-9
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00273370
InChI Key IOGISYQVOGVIEU-VKHMYHEASA-N
Synonym s-4-hydroxy-2-pyrrolidinone, s-4-hydroxypyrrolidin-2-one, s-4-hydroxy-2-pyrrolidone, s-4-hydroxypyrrolidine-2-one, 4s-4-hydroxypyrrolidin-2-one, s---4-hydroxy-2-pyrrolidinone, 2-pyrrolidinone, 4-hydroxy-, 4s, s---4-hydroxy-2-pyrrolidone, s-4-hydroxy-pyrrolidine-2-one, 4s-4-hydroxy-2-pyrrolidinone
PubChem CID 155084
IUPAC Name (4S)-4-hydroxypyrrolidin-2-one
SMILES C1C(CNC1=O)O

Specifications

Melting Point 157°C
Quantity 1 g
Formula Weight 101.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidone
EINECSNumber (5)-5886
TSCA No