missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-Hydroxy-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Catalog No. AC398970050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC398970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC398970050 Supplier Thermo Scientific Chemicals Supplier No. 398970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68108-18-9 | Molecular Formula: C4H7NO2 | Molecular Weight: 101.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68108-18-9
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.1
InChI Key IOGISYQVOGVIEU-VKHMYHEASA-N
Synonym s-4-hydroxy-2-pyrrolidinone, s-4-hydroxypyrrolidin-2-one, s-4-hydroxy-2-pyrrolidone, s-4-hydroxypyrrolidine-2-one, 4s-4-hydroxypyrrolidin-2-one, s---4-hydroxy-2-pyrrolidinone, 2-pyrrolidinone, 4-hydroxy-, 4s, s---4-hydroxy-2-pyrrolidone, s-4-hydroxy-pyrrolidine-2-one, 4s-4-hydroxy-2-pyrrolidinone
PubChem CID 155084
IUPAC Name (4S)-4-hydroxypyrrolidin-2-one
SMILES C1C(CNC1=O)O

Specifications

Melting Point 156°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 101.1
Percent Purity 98%
Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only