missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyl-2-oxazolidinone 98.0+%, TCI America™

Supplier:  TCI America B175425G

Quantity 25 g

 View more versions of this product

Catalog No. B175425G


Only null left

Description

Description

Specifications

Chemical Identifiers

90719-32-7
177.20
OJOFMLDBXPDXLQ-VIFPVBQESA-N
736225
O=C1N[C@@H](CC2=CC=CC=C2)CO1
C10H11NO2
MFCD00064496
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
(4S)-4-benzyl-1,3-oxazolidin-2-one

Specifications

88°C
90719-32-7
C10H11NO2
s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
O=C1N[C@@H](CC2=CC=CC=C2)CO1
25 g
736225
≥98.0% (GC)
(S)-4-Benzyl-2-oxazolidinone
White-Yellow
MFCD00064496
OJOFMLDBXPDXLQ-VIFPVBQESA-N
(4S)-4-benzyl-1,3-oxazolidin-2-one
177.20
177.20
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions