missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-(4-Aminobenzyl)-2-oxazolidinone 98.0+%, TCI America™
$31.68 - $95.06
Chemical Identifiers
| CAS | 152305-23-2 |
|---|---|
| Molecular Formula | C10H12N2O2 |
| Molecular Weight (g/mol) | 192.218 |
| MDL Number | MFCD03411476 |
| InChI Key | WNAVSKJKDPLWBD-VIFPVBQESA-N |
| Synonym | s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one |
| PubChem CID | 7099156 |
| IUPAC Name | (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one |
| SMILES | C1C(NC(=O)O1)CC2=CC=C(C=C2)N |
Chemical Identifiers
| 152305-23-2 | |
| 192.218 | |
| WNAVSKJKDPLWBD-VIFPVBQESA-N | |
| 7099156 | |
| C1C(NC(=O)O1)CC2=CC=C(C=C2)N |
| C10H12N2O2 | |
| MFCD03411476 | |
| s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one | |
| (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one |
Specifications
| 152305-23-2 | |
| C10H12N2O2 | |
| 1 g | |
| WNAVSKJKDPLWBD-VIFPVBQESA-N | |
| (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one | |
| 7099156 | |
| ≥98.0% (GC,T) | |
| (S)-4-(4-Aminobenzyl)-2-oxazolidinone |
| 110°C | |
| MFCD03411476 | |
| s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one | |
| C1C(NC(=O)O1)CC2=CC=C(C=C2)N | |
| 192.218 | |
| 192.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No