missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(4-Aminobenzyl)-2-oxazolidinone 98.0+%, TCI America™

Catalog No. A20951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20951G 1 g
A20955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20951G Supplier TCI America Supplier No. A20951G
Only null left

Chemical Identifiers

CAS 152305-23-2
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD03411476
InChI Key WNAVSKJKDPLWBD-VIFPVBQESA-N
Synonym s-4-4-aminobenzyl oxazolidin-2-one, s-4-4-aminobenzyl-2-oxazolidinone, s-4-4-aminobenzyl-2 1h-oxazolidinone, s-4-4-aminobenzyl-1,3-oxazolidin-2-one, s-4-4-amino-benzyl-oxazolidine-2-one, 4s-4-4-aminophenyl methyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-4-aminophenyl methyl-, 4s, s-4-4-aminobenzyl-1,3-oxozolidin-2-one, 4s-4-4-aminophenyl methyl-2-oxazolidinone, s-4-4'-aminobenzyl-1,3-oxazolidin-2-one
PubChem CID 7099156
IUPAC Name (4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
SMILES C1C(NC(=O)O1)CC2=CC=C(C=C2)N

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 192.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-(4-Aminobenzyl)-2-oxazolidinone