missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee)

Catalog No. AC393372500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393372500 250 mg Glass bottle
AC393370010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393372500 Supplier Thermo Scientific Chemicals Supplier No. 393372500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54656-96-1 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54656-96-1
MDL Number MFCD00077866
InChI Key HVOAMIOKNARIMR-LURJTMIESA-N
Synonym s-1-pyridin-4-yl ethanol, s---1-4-pyridyl ethanol, 1s-1-pyridin-4-yl ethan-1-ol, 1s-1-pyridin-4-yl ethanol, s-1-4-pyridyl ethanol, 1s-1-4-pyridyl ethanol, s-1-4-pyridinyl ethanol, s--1-4-pyridyl ethanol, 1s-1-4-pyridinyl ethanol, --s-1-4-pyridyl ethanol
PubChem CID 10920507
IUPAC Name (1S)-1-pyridin-4-ylethanol
SMILES CC(C1=CC=NC=C1)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 67°C to 70°C
Color White
Enantiomeric Excess 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C7H9NO
Quantity 250 mg
Specific Rotation Condition −58.50° (20°C c=1,chloroform)
Specific Rotation -56° to -61° (20°C, 589nm) (C=1, chloroform)
Molecular Weight (g/mol) 123.155
Formula Weight 123.15
Percent Purity ≥99.0%
Physical Form Needles
Chemical Name or Material (S)-4-(1-Hydroxyethyl)pyridine
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only