missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-tert-Butylamino-1,2-propanediol 96.0+%, TCI America™

Catalog No. B19875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19875G Supplier TCI America Supplier No. B19875G
Only null left

Chemical Identifiers

CAS 30315-46-9
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00190165
InChI Key JWBMVCAZXJMSOX-LURJTMIESA-N
PubChem CID 2733671
IUPAC Name (2S)-3-(tert-butylamino)propane-1,2-diol
SMILES CC(C)(C)NCC(CO)O

Specifications

Melting Point 81°C
Quantity 5 g
Formula Weight 147.22
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-3-tert-Butylamino-1,2-propanediol