missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(tert-Butoxycarbonylamino)piperidine 98.0+%, TCI America™

Catalog No. p-7123083
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36601G 1 g
B36605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36601G Supplier TCI America Supplier No. B36601G

Chemical Identifiers

CAS 216854-23-8
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093383
InChI Key WUOQXNWMYLFAHT-QMMMGPOBSA-N
Synonym s-3-boc-aminopiperidine, s-3-n-boc-aminopiperidine, s-tert-butyl piperidin-3-ylcarbamate, s-3-boc-amino piperidine, tert-butyl n-3s-piperidin-3-yl carbamate, s-3-tert-butoxycarbonylamino piperidine, tert-butyl s-piperidin-3-ylcarbamate, s-3-n-boc-amino piperidine, s---3-tert-butoxycarbonylaminopiperidine, 3s-3-aminopiperidine, 3-boc protected
PubChem CID 1514171
IUPAC Name tert-butyl N-[(3S)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-(tert-Butoxycarbonylamino)piperidine