missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95.0+%, TCI America™

Catalog No. B17591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17591G 1 g
B17595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17591G Supplier TCI America Supplier No. B17591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102308-32-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00209557
InChI Key PNJXYVJNOCLJLJ-MRVPVSSYSA-N
Synonym s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
PubChem CID 179824
IUPAC Name tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C

Specifications

Color Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine
TSCA No
Recommended Storage Refrigerator