missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3-Piperidinecarboxylic Acid 98.0+%, TCI America™
$55.84 - $175.20
Chemical Identifiers
| CAS | 59045-82-8 |
|---|---|
| Molecular Formula | C6H11NO2 |
| Molecular Weight (g/mol) | 129.159 |
| MDL Number | MFCD01630807 |
| InChI Key | XJLSEXAGTJCILF-YFKPBYRVSA-N |
| Synonym | s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid |
| PubChem CID | 6575389 |
| ChEBI | CHEBI:222169 |
| IUPAC Name | (3S)-piperidine-3-carboxylic acid |
| SMILES | C1CC(CNC1)C(=O)O |
Chemical Identifiers
| 59045-82-8 | |
| 129.159 | |
| XJLSEXAGTJCILF-YFKPBYRVSA-N | |
| 6575389 | |
| (3S)-piperidine-3-carboxylic acid |
| C6H11NO2 | |
| MFCD01630807 | |
| s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid | |
| CHEBI:222169 | |
| C1CC(CNC1)C(=O)O |
Specifications
| 59045-82-8 | |
| C6H11NO2 | |
| s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid | |
| C1CC(CNC1)C(=O)O | |
| 1 g | |
| 6575389 | |
| 129.16 | |
| Crystalline Powder |
| White | |
| MFCD01630807 | |
| XJLSEXAGTJCILF-YFKPBYRVSA-N | |
| (3S)-piperidine-3-carboxylic acid | |
| 129.159 | |
| CHEBI:222169 | |
| ≥98.0% (T) | |
| (S)-(+)-3-Piperidinecarboxylic Acid |
Safety and Handling
TSCA : No