missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. N06785G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06785G 5 g
N06781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06785G Supplier TCI America Supplier No. N06785G

Chemical Identifiers

CAS 59045-82-8
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD01630807
InChI Key XJLSEXAGTJCILF-YFKPBYRVSA-N
Synonym s-piperidine-3-carboxylic acid, s-+-nipecotic acid, s-nipecotic acid, 3s-piperidine-3-carboxylic acid, +-nipecotic acid, s-+-3-piperidinecarboxylic acid, 3s-nipecotic acid, l +-nipecotic acid, s-hexahydronicotinic acid, +-hexahydronicotinic acid
PubChem CID 6575389
ChEBI CHEBI:222169
IUPAC Name (3S)-piperidine-3-carboxylic acid
SMILES C1CC(CNC1)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-3-Piperidinecarboxylic Acid