missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Methylmorpholine Hydrochloride 98.0+%, TCI America™
$246.05 - $941.35
Chemical Identifiers
| CAS | 1022094-03-6 |
|---|---|
| Molecular Formula | C5H12ClNO |
| Molecular Weight (g/mol) | 137.607 |
| MDL Number | MFCD18382512 |
| InChI Key | MSOCQCWIEBVSLF-JEDNCBNOSA-N |
| Synonym | s-3-methylmorpholine hydrochloride, s-3-methylmorpholine hcl, 3s-3-methylmorpholine hydrochloride, s-3-methyl-morpholine hydrochloride, s-3-methyl morpholine hydrochloride, c5h11no.clh, s-3-methylmorpholinehydrochloride, s-3-methylmorpholin hydrochloride, 3s-3-methylmorpholine-hydrogen chloride 1/1 |
| PubChem CID | 53486469 |
| IUPAC Name | (3S)-3-methylmorpholine;hydrochloride |
| SMILES | CC1COCCN1.Cl |
Chemical Identifiers
| 1022094-03-6 | |
| 137.607 | |
| MSOCQCWIEBVSLF-JEDNCBNOSA-N | |
| 53486469 | |
| CC1COCCN1.Cl |
| C5H12ClNO | |
| MFCD18382512 | |
| s-3-methylmorpholine hydrochloride, s-3-methylmorpholine hcl, 3s-3-methylmorpholine hydrochloride, s-3-methyl-morpholine hydrochloride, s-3-methyl morpholine hydrochloride, c5h11no.clh, s-3-methylmorpholinehydrochloride, s-3-methylmorpholin hydrochloride, 3s-3-methylmorpholine-hydrogen chloride 1/1 | |
| (3S)-3-methylmorpholine;hydrochloride |
Specifications
| 1022094-03-6 | |
| White | |
| MFCD18382512 | |
| s-3-methylmorpholine hydrochloride, s-3-methylmorpholine hcl, 3s-3-methylmorpholine hydrochloride, s-3-methyl-morpholine hydrochloride, s-3-methyl morpholine hydrochloride, c5h11no.clh, s-3-methylmorpholinehydrochloride, s-3-methylmorpholin hydrochloride, 3s-3-methylmorpholine-hydrogen chloride 1/1 | |
| CC1COCCN1.Cl | |
| 137.607 | |
| 137.61 | |
| Crystalline Powder |
| 128°C | |
| C5H12ClNO | |
| 1 g | |
| MSOCQCWIEBVSLF-JEDNCBNOSA-N | |
| (3S)-3-methylmorpholine;hydrochloride | |
| 53486469 | |
| ≥98.0% (T) | |
| (S)-3-Methylmorpholine Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator