missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3-Methyl-1-pentanol 98.0+%, TCI America™
Supplier: TCI America M09635ML
Specifications
| (S)-(+)-3-Methyl-1-pentanol | |
| Colorless | |
| C6H14O | |
| 5 mL | |
| IWTBVKIGCDZRPL-UHFFFAOYNA-N | |
| 3-methylpentan-1-ol | |
| 641003 | |
| ≥98.0% (GC) |
| 42072-39-9 | |
| 152°C | |
| MFCD00135167 | |
| 1987 | |
| CCC(C)CCO | |
| 102.18 | |
| 102.18 | |
| Liquid |
Chemical Identifiers
| 42072-39-9 | |
| 102.18 | |
| IWTBVKIGCDZRPL-UHFFFAOYNA-N | |
| 3-methylpentan-1-ol |
| C6H14O | |
| MFCD00135167 | |
| 641003 | |
| CCC(C)CCO |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No