missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Hydroxytetrahydrofuran, 99%

Catalog No. AC304430050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304430050 5 g Glass bottle
AC304430250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304430050 Supplier Thermo Scientific Chemicals Supplier No. 304430050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86087-23-2 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86087-23-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00064327
InChI Key XDPCNPCKDGQBAN-BYPYZUCNSA-N
Synonym s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s
PubChem CID 2733227
IUPAC Name (3S)-oxolan-3-ol
SMILES C1COCC1O

Specifications

Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Density 1.2980g/mL
Quantity 5 g
Boiling Point 80.0°C (15.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4490 to 1.4510
Specific Gravity 1.298
Assay Percent Range 98.5% min. (GC)
Specific Rotation Condition +18.5° (20°C c=2 MeOH)
Specific Rotation + 18.50
Solubility Information Solubility in water: soluble in water.
Formula Weight 88.11
Percent Purity 99%
Chemical Name or Material (S)-(+)-3-Hydroxytetrahydrofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only