missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Hydroxytetrahydrofuran, 98%

Catalog No. AAH5647606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5647606 5 g
AAH5647614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5647606 Supplier Thermo Scientific Chemicals Supplier No. H5647606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86087-23-2 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86087-23-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00064327
InChI Key XDPCNPCKDGQBAN-BYPYZUCNSA-N
Synonym s-+-3-hydroxytetrahydrofuran, s-tetrahydrofuran-3-ol, s-3-hydroxytetrahydrofuran, 3s-oxolan-3-ol, s-+-tetrahydro-3-furanol, s-tetrahydro-3-furanol, s-+-tetrahyro-3-furanol, 3s-tetrahydrofuran-3-ol, s-tetrahydro-furan-3-ol, 3-furanol, tetrahydro-, 3s
PubChem CID 2733227
IUPAC Name (3S)-oxolan-3-ol
SMILES C1COCC1O

Specifications

Density 1.103
Quantity 5 g
Refractive Index 1.45
Beilstein 4652609
Boiling Point 80°C (15 mmHg)
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-3-Hydroxytetrahydrofuran
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only