missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxypyrrolidine hydrochloride, 97%

Catalog No. AAH5281506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5281506 5 g
AAH5281503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5281506 Supplier Thermo Scientific Chemicals Supplier No. H5281506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 122536-94-1 | C4H10ClNO | 123.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122536-94-1
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
MDL Number MFCD00272298
InChI Key QPMSJEFZULFYTB-WCCKRBBISA-N
Synonym s-3-hydroxypyrrolidine hydrochloride, s-pyrrolidin-3-ol hydrochloride, 3s-pyrrolidin-3-ol hydrochloride, s-+-3-pyrrolidinol hydrochloride, s-3-hydroxypyrrolidine-hcl, s-3-pyrrolidinol hydrochloride, 3s-pyrrolidinol hydrochloride, s-3-pyrrolidinol hcl, s-pyrrolidin-3-ol, hcl, s---3-pyrrolidinol hydrochloride
PubChem CID 22309122
IUPAC Name (3S)-pyrrolidin-3-ol;hydrochloride
SMILES C1CNCC1O.Cl

Specifications

Melting Point 106°C to 107°C
Boiling Point 108°C to 110°C (8 mmHg)
Quantity 5 g
Formula Weight 123.58
Percent Purity 97%
Chemical Name or Material (S)-3-Hydroxypyrrolidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only