missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Hydroxy-3-phenylpropanoic acid, 99%

Catalog No. p-3778853
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412120010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412120010 Supplier Thermo Scientific Chemicals Supplier No. 412120010
Only null left

CAS: 36567-72-3 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36567-72-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00145262
InChI Key AYOLELPCNDVZKZ-QMMMGPOBSA-N
Synonym s-3-hydroxy-3-phenylpropanoic acid, 3s-3-hydroxy-3-phenylpropanoic acid, s-3-hydroxy-3-phenylpropionic acid, ?-3-phenylhydracrylic acid, s---3-hydroxy-3-phenylpropionic acid, benzenepropanoic acid,, a-hydroxy-,, as, s---3-hydroxy-3-phenylpropionic acid, 98+%, unii-s2w4lws09c component, s-3-hydroxy-3-phenylpropionic acid sum of enantiomers, gc
PubChem CID 2735057
ChEBI CHEBI:51058
IUPAC Name (3S)-3-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 116.0°C to 119.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −22.50° (20°C c=4,CH3OH)
Specific Rotation − 22.50
Formula Weight 166.18
Percent Purity 99%
Physical Form Needle-like Crystals
Chemical Name or Material (S)-3-Hydroxy-3-phenylpropanoic acid
Show More Show Less

RUO – Research Use Only