missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Fluoropyrrolidine hydrochloride, 97%

Catalog No. AC442740010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442740010 1 g
AC442740050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442740010 Supplier Thermo Scientific Chemicals Supplier No. 442740010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 136725-53-6 | C4H8FN·ClH | 125.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136725-53-6
Molecular Formula C4H8FN·ClH
Molecular Weight (g/mol) 125.57
InChI Key LENYOXXELREKGZ-WCCKRBBISA-N
Synonym s-3-fluoropyrrolidine hydrochloride, s-+-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine hydrochloride, 3s-+-3-fluoropyrrolidine hydrochloride, s-3-fluoro-pyrrolidine hcl, s ;-3-fluoropyrrolidine hydrochloride, 3s-3-fluoropyrrolidine, chloride, s-3-fluoropyrrolidine hcl
PubChem CID 16217739
IUPAC Name (3S)-3-fluoropyrrolidine;hydrochloride
SMILES C1CNCC1F.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180°C to 182°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 6.15
Formula Weight 125.57
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material (S)-3-Fluoropyrrolidine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only