missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(Dimethylamino)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog No. p-4812519
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426450010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426450010 Supplier Thermo Scientific Chemicals Supplier No. 426450010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132883-44-4
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
InChI Key AVAWMINJNRAQFS-LURJTMIESA-N
Synonym 3s---3-dimethylamino pyrrolidine, s-3-dimethylaminopyrrolidine, s---3-dimethylamino pyrrolidine, s-n,n-dimethylpyrrolidin-3-amine, 3s-n,n-dimethylpyrrolidin-3-amine, s-3-dimethylamino pyrrolidine, 3s-n,n-dimethyl-3-pyrrolidinamine, 3s-3-dimethylaminopyrrolidine, 3-pyrrolidinamine, n,n-dimethyl-, 3s, s-dimethyl-pyrrolidin-3-yl-amine
PubChem CID 7019156
IUPAC Name (3S)-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

Density 0.8990g/mL
Boiling Point 164.0°C
Flash Point 51°C
Assay Percent Range 96% (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.899
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material (S)-(-)-3-(Dimethylamino)pyrrolidine
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only