missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™

$42.36 - $119.36

Chemical Identifiers

CAS 132335-44-5
Molecular Formula C9H15NOS
Molecular Weight (g/mol) 185.29
MDL Number MFCD07782107
InChI Key XWCNSHMHUZCRLN-UHFFFAOYNA-N
Synonym (S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene
PubChem CID 9964277
IUPAC Name 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
SMILES CN(C)CCC(O)C1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D40101G
View Documents
TCI America
D40101G
1 g
Each for $42.36
Only null left
 
D40105G
View Documents
TCI America
D40105G
5 g
Each for $119.36
Only null left
 
Description

Description

Specifications

Chemical Identifiers

132335-44-5
185.29
XWCNSHMHUZCRLN-UHFFFAOYNA-N
9964277
CN(C)CCC(O)C1=CC=CS1
C9H15NOS
MFCD07782107
(S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene
3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol

Specifications

132335-44-5
White
MFCD07782107
(S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene
CN(C)CCC(O)C1=CC=CS1
185.29
185.29
Crystalline Powder
79°C
C9H15NOS
1 g
XWCNSHMHUZCRLN-UHFFFAOYNA-N
3-(dimethylamino)-1-(thiophen-2-yl)propan-1-ol
9964277
≥98.0% (GC,T)
(S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'