missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Chloro-1,2-propanediol 98.0+%, TCI America™

Catalog No. C154225G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C154225G 25 g
C154210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C154225G Supplier TCI America Supplier No. C154225G
Only null left

Chemical Identifiers

CAS 60827-45-4
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00210270
InChI Key SSZWWUDQMAHNAQ-GSVOUGTGSA-N
Synonym s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
PubChem CID 148904
IUPAC Name (2S)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O

Specifications

Boiling Point 80°C
Quantity 25 g
UN Number 2689
Formula Weight 110.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-3-Chloro-1,2-propanediol
EINECSNumber (2)-2441
RTECSNumber TY4202300
TSCA No