missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3-Chloro-1,2-propanediol 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 80°C |
| Quantity | 10 g |
| UN Number | 2689 |
| Formula Weight | 110.54 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-(+)-3-Chloro-1,2-propanediol |
Safety and Handling
| EINECSNumber | (2)-2441 |
| RTECSNumber | TY4202300 |
| TSCA | No |