missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%

CAS: 60827-45-4 | C3H7ClO2 | 110.537 g/mol

$134.16 - $448.38

Chemical Identifiers

CAS 60827-45-4
Molecular Formula C3H7ClO2
Molecular Weight (g/mol) 110.537
MDL Number MFCD00210270
InChI Key SSZWWUDQMAHNAQ-GSVOUGTGSA-N
Synonym s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
PubChem CID 148904
IUPAC Name (2S)-3-chloropropane-1,2-diol
SMILES C(C(CCl)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1855606
View Documents
Thermo Scientific Chemicals
L1855606
5 g
Each for $134.16
Only null left
 
AAL1855614
View Documents
Thermo Scientific Chemicals
L1855614
25 g
Each for $448.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60827-45-4
110.537
SSZWWUDQMAHNAQ-GSVOUGTGSA-N
148904
C(C(CCl)O)O
C3H7ClO2
MFCD00210270
s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
(2S)-3-chloropropane-1,2-diol

Specifications

60827-45-4
213°C
C3H7ClO2
MFCD00210270
UN2689
s-+-3-chloro-1,2-propanediol, s-3-chloro-1,2-propanediol, s-3-chloropropane-1,2-diol, 2s-3-chloropropane-1,2-diol, s-alpha-chlorohydrin, unii-227bd3h6h6, ccris 7386, 1,2-propanediol, 3-chloro-, s, 1,2-propanediol, 3-chloro-, 2s
+0.9° (Neat)
(2S)-3-chloropropane-1,2-diol
148904
98%
ee 98%
1.322
126°C (258°F)
1.48
5 g
2202929
SSZWWUDQMAHNAQ-GSVOUGTGSA-N
C(C(CCl)O)O
110.537
110.54
(S)-(+)-3-Chloro-1,2-propanediol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H360-H341-H318-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Causes serious eye damage.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H318-H330-H335-H341-H360

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN

RTECSNumber : TY4202300

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only