missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-3-Chloro-1,2-propanediol, 98%, ee 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.322 |
| Boiling Point | 213°C |
| Flash Point | 126°C (258°F) |
| Refractive Index | 1.48 |
| Quantity | 25 g |
| UN Number | UN2689 |
| Beilstein | 2202929 |
| Optical Rotation | +0.9° (Neat) |
| Formula Weight | 110.54 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H301-H312-H315-H318-H330-H335-H341-H360 |
| Hazard Statement | GHS H Statement H301-H331-H360-H341-H318-H312-H315-H335 Toxic if swallowed. Toxic if inhaled. May damage fertility or the unborn child. Suspected of causing genetic defects. Causes serious eye damage. Harmful in contact with skin. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: GLYCEROL ALPHA-MONOCHLOROHYDRIN |
| RTECSNumber | TY4202300 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only